missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Dnmt2, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen This antibody was raised against synthetic peptides corresponding amino acids 39-53 and 361-376 of mouse Dnmt2.
Antigen Dnmt2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB200616 Novus Biologicals Canada
NB200616
0.1mg Each for $309.05
NB200616SS Novus Biologicals Canada
NB200616SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
Dnmt2
Purified
RUO
Cross-reacts with Human and Mouse. Not yet tested in other species.
PBS containing 0.05% BSA
This antibody was raised against synthetic peptides corresponding amino acids 39-53 and 361-376 of mouse Dnmt2.
Polyclonal
Primary
Western Blot
1787
0.5mg/mL