Please login to your online account to display your discounted pricing

anti-DNMT1, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen Contained within amino acids 1-125 of the N-terminus of human Dnmt1 [UniProt No. P26358]
Antigen DNMT1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100264SS

 
novus biologicals canada
NB100264SS
Unlabeled 0.025mL Each for $144.63

NB100264

 
novus biologicals canada
NB100264
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Contained within amino acids 1-125 of the N-terminus of human Dnmt1 [UniProt No. P26358]
Antigen DNMT1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Gene Accession No. P26358
Gene ID (Entrez) 1786