Please login to your online account to display your discounted pricing

anti-DNMT1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100264

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100264

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1786
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Format Antiserum
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Primary or Secondary Primary
Quantity 0.1mL
Gene Accession No. P26358
Host Species Rabbit
Antigen DNMT1
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen Contained within amino acids 1-125 of the N-terminus of human Dnmt1 [UniProt No. P26358]
Regulatory Status RUO