Please login to your online account to display your discounted pricing

anti-DMAP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203982

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203982

  • / Each
 

Description & Specifications

Specifications

Antigen DMAP1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. NP_001029196
Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human DMAP1 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55929
Test Specificity Reacts with Chimpanzee, Cow, Dog, Human, Mouse and Rat. Predicted to react with Chicken and Zebrafish. The amino acid sequence used as immunogen is 100% homologous in Chimpanzee, Dog, Human, Rat, Cow, Mouse, 88% homologous in Chicken and 77% homologous in Zebrafish.
Target Species Bovine
Target Species Canine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat