missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-DMAP1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human DMAP1 was used as the immunogen.
Antigen DMAP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP203982SS Novus Biologicals Canada
NBP203982SS
0.025mg Each for $139.00
NBP203982 Novus Biologicals Canada
NBP203982
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
DMAP1
Purified
RUO
Reacts with Chimpanzee, Cow, Dog, Human, Mouse and Rat. Predicted to react with Chicken and Zebrafish. The amino acid sequence used as immunogen is 100% homologous in Chimpanzee, Dog, Human, Rat, Cow, Mouse, 88% homologous in Chicken and 77% homologous in Zebrafish.
55929
0.5mg/mL
A portion of amino acids 250-300 of human DMAP1 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
NP_001029196
100μg in 200μL PBS containing 0.05% BSA