Please login to your online account to display your discounted pricing

anti-DLL4, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 550-610 of human DLL4 was used as the immunogen. (aa561-577)
Antigen DLL4
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227407

 
novus biologicals canada
NBP227407
Unlabeled 0.1mg Each for $307.00

NBP227407SS

 
novus biologicals canada
NBP227407SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 550-610 of human DLL4 was used as the immunogen. (aa561-577)
Antigen DLL4
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 94% homologous in mouse.
Gene Accession No. Q9NR61
Gene ID (Entrez) 54567
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL