Please login to your online account to display your discounted pricing

anti-DLL1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227088SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227088SS

  • / Each
 

Description & Specifications

Specifications

Antigen DLL1
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Gene Accession No. O00548
Host Species Rabbit
Immunogen A portion of amino acids 640-728 of human DLL1 was used as the immunogen for the antibody. (aa649-665)
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 28514
Test Specificity The amino acid sequence used as immunogen for the DLL1 antibody is 100% homologous in human and chimp, 82% homologous in rhesus monkey, horse and rat, and 76% homologous in mouse.
Target Species Human
Target Species Primate