Please login to your online account to display your discounted pricing

anti-DLL1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227088

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227088

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 28514
Gene Accession No. O00548
Monoclonal or Polyclonal Polyclonal
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Test Specificity The amino acid sequence used as immunogen for the DLL1 antibody is 100% homologous in human and chimp, 82% homologous in rhesus monkey, horse and rat, and 76% homologous in mouse.
Concentration 0.5mg/mL
Regulatory Status RUO
Conjugate Unlabeled
Antigen DLL1
Cross Reactivity Human
Cross Reactivity Primate
Immunogen A portion of amino acids 640-728 of human DLL1 was used as the immunogen for the antibody. (aa649-665)
Quantity 0.1mg
Host Species Rabbit
Primary or Secondary Primary
Applications Immunohistochemistry (Paraffin)