missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-DLL1, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 640-728 of human DLL1 was used as the immunogen for the antibody. (aa649-665)
Antigen DLL1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227088 Novus Biologicals Canada
NBP227088
0.1mg Each for $307.00
NBP227088SS Novus Biologicals Canada
NBP227088SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
DLL1
Purified
RUO
The amino acid sequence used as immunogen for the DLL1 antibody is 100% homologous in human and chimp, 82% homologous in rhesus monkey, horse and rat, and 76% homologous in mouse.
28514
0.5mg/mL
A portion of amino acids 640-728 of human DLL1 was used as the immunogen for the antibody. (aa649-665)
Polyclonal
Primary
Immunohistochemistry (Paraffin)
O00548
100μg in 200μL PBS containing 0.05% BSA.