Please login to your online account to display your discounted pricing

Dkk-4 Mouse anti-Human, Clone: 185709, R&D Systems


Mouse Monoclonal Antibody

Manufacturer: r & d systems  RDSMAB1269500

 View more versions of this product

Catalog No. MAB1269500

  • / Each
 

Description & Specifications

Specifications

Clone 185709
Regulatory Status RUO
Cross Reactivity Human
Antigen Dkk-4
Isotype IgG2b
Format Purified
Host Species Mouse
Quantity 500μg
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Immunogen Mouse myeloma cell line NS0-derived recombinant human Dkk-4. Leu19-Leu224
Purification Method Protein A or G purified from hybridoma culture supernatant

Dkk-4 Monoclonal antibody MAB1269500 Specifically detects Dkk-4 in Human samples. It is validated for ELISA Capture, Western Blotting