Please login to your online account to display your discounted pricing

Dkk-3 Rat anti-Mouse, Clone: 153103, R&D Systems

Rat Monoclonal Antibody

$207.26 - $441.90

Specifications

Antigen Dkk-3
Clone 153103
Cross Reactivity Mouse
Format Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB948050

 
r & d systems
RDSMAB948050
Unlabeled 50μg Each for $207.26

MAB948500

 
r & d systems
RDSMAB948500
Unlabeled 500μg Each for $441.90
Description & Specifications

Specifications

Antigen Dkk-3
Clone 153103
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Dkk-3. Ala22-Ile349
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Dkk-3 Monoclonal antibody MAB948 Specifically detects Dkk-3 in Mouse samples. It is validated for Western Blotting