Please login to your online account to display your discounted pricing

anti-Dishevelled-3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224674SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224674SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1857
Formulation 100μg in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and cow, 89% homologous in mouse and rat, and 84% homologous in chicken.
Gene Accession No. Q92997
Primary or Secondary Primary
Concentration 0.5mg/mL
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Regulatory Status RUO
Format Purified
Immunogen A portion of amino acids 560-610 of human DVL-3 was used as the immunogen.
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Rabbit
Antigen Dishevelled-3
Quantity 0.025mg