Please login to your online account to display your discounted pricing

anti-Dishevelled-3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224674

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224674

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1857
Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Gene Accession No. Q92997
Quantity 0.1mg
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and cow, 89% homologous in mouse and rat, and 84% homologous in chicken.
Concentration 0.5mg/mL
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Regulatory Status RUO
Immunogen A portion of amino acids 560-610 of human DVL-3 was used as the immunogen.
Antigen Dishevelled-3