Please login to your online account to display your discounted pricing

anti-Dishevelled-3, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 560-610 of human DVL-3 was used as the immunogen.
Antigen Dishevelled-3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224674SS

 
novus biologicals canada
NBP224674SS
Unlabeled 0.025mg Each for $139.00

NBP224674

 
novus biologicals canada
NBP224674
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 560-610 of human DVL-3 was used as the immunogen.
Antigen Dishevelled-3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and cow, 89% homologous in mouse and rat, and 84% homologous in chicken.
Gene Accession No. Q92997
Gene ID (Entrez) 1857
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL