Please login to your online account to display your discounted pricing

anti-Dishevelled-1, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 70-120 of human DVL-1 was used as the immunogen.
Antigen Dishevelled-1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224648

 
novus biologicals canada
NBP224648
Unlabeled 0.1mg Each for $307.00

NBP224648SS

 
novus biologicals canada
NBP224648SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 70-120 of human DVL-1 was used as the immunogen.
Antigen Dishevelled-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, pig, opossum, chimpanzee, dog and chicken, and 94% homologous in rat and mouse.
Gene Accession No. O14640
Gene ID (Entrez) 1855
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL