Please login to your online account to display your discounted pricing

anti-DISC1, Polyclonal, Novus Biologicals™

$144.63 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 400-500) of the rat DISC1 protein. [Swiss-Prot No. Q810H6]
Antigen DISC1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11040773

 
novus biologicals canada
NB11040773
Unlabeled 0.1mL Each for $309.05

NB11040773T

 
novus biologicals canada
NB11040773SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 400-500) of the rat DISC1 protein. [Swiss-Prot No. Q810H6]
Antigen DISC1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Gene ID (Entrez) 27185
Formulation Tris-glycine, 150mM NaCl
Concentration 0.85mg/mL