missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-DIO3, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat DIO3 (within residues 1-100). [Swiss-Prot No. P49897]
Antigen DIO3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11096414T Novus Biologicals Canada
NB11096414SS
0.025mL Each for $139.00
NB11096414 Novus Biologicals Canada
NB11096414
0.1mL Each for $329.00
Specifications Description & Specifications

Specifications

Rabbit
DIO3
Affinity Purified
RUO
Human, rat, mouse and hamster. Human reactivity was confirmed in PMID 22723689.
PBS, 30% glycerol
Synthetic peptide made to an internal portion of rat DIO3 (within residues 1-100). [Swiss-Prot No. P49897]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Western Blot
1735
1mg/mL