Please login to your online account to display your discounted pricing

anti-DIO3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11096414

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11096414

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1735
Formulation PBS, 30% glycerol
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Regulatory Status RUO
Format Affinity Purified
Primary or Secondary Primary
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Conjugate Unlabeled
Test Specificity Human, rat, mouse and hamster. Human reactivity was confirmed in PMID 22723689.
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen Synthetic peptide made to an internal portion of rat DIO3 (within residues 1-100). [Swiss-Prot No. P49897]
Quantity 0.1mL
Antigen DIO3