missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl Histone H4 (Arg3), Asymmetric Rabbit anti-Human, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$118.34 - $453.63

Specifications

Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4, asymmetrically dimethylated at Arg3.
Antigen dimethyl Histone H4 (Arg3), Asymmetric
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ABE441SMI View Documents MilliporeSigma
ABE441S
10μL Each for $118.34
 
ABE441MI View Documents MilliporeSigma
ABE441
100μL Each for $453.63
 
Description

Description

This Anti-dimethyl Histone H4 (Arg3) antibody, Asymmetric is validated for use in western blotting, immunocytochemistry and dot blot for the detection of dimethyl Histone H4 (Arg3).
Specifications

Specifications

Rabbit
dimethyl Histone H4 (Arg3), Asymmetric
Affinity Purified
Dot Blot, Immunocytochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4, asymmetrically dimethylated at Arg3.
Polyclonal
Primary
NP_001029249

For Research Use Only