Dimethyl-Histone H4 (Arg3) Asymmetric (Trial Size) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07213I
Description
This Anti-dimethyl-Histone H4 (Arg3) Asymmetric Antibody is validated for use in Western Blotting and Immunoprecipitation and Dot Blot for the detection of dimethyl-Histone H4 (Arg3) Asymmetrimmunocytochemistry.Specifications
dimethyl-Histone H4 (Arg3) Asymmetric | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Dot Blot, Immunoprecipitation, Western Blot | |
Unconjugated | |
HIST1H4A; H4/A; H4FA; HIST1H4B; H4/I; H4FI; HIST1H4C; H4/G; H4FG; HIST1H4D; H4/B; H4FB; HIST1H4E; H4/J; H4FJ; HIST1H4F; H4/C; H4FC; HIST1H4H; H4/H; H4FH; HIST1H4I; H4/M; H4FM; HIST1H4J; H4/E; H4FE; HIST1H4K; H4/D; H4FD; HIST1H4L; H4/K; H4FK; HIST2H4A; H4/N; H4F2; H4FN; HIST2H4; HIST2H4B; H4/O; | |
KLH-conjugated linear peptide corresponding to the N-terminus of human dimethyl-Histone H4 (Arg3) Asymmetric. | |
100 μL | |
NP_001029249 | |
2-8°C for 12 months |
For Research Use Only