Please login to your online account to display your discounted pricing

anti-dimethyl Histone H4 (Arg3), Asymmetric, Polyclonal, EMD Millipore™


Manufacturer: emd millipore corporation  ABE441S

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. ABE441SMI

  • / Each
 

Description & Specifications

Specifications

Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Quantity 10μL
Primary or Secondary Prrimary
Cross Reactivity Human
Antigen dimethyl Histone H4 (Arg3), Asymmetric
Host Species Rabbit
Applications Dot Blot
Applications Immunocytochemistry
Applications Western Blotting
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene Symbols HIST1H4A; H4/A; H4FA; HIST1H4B; H4/I; H4FI; HIST1H4C; H4/G; H4FG; HIST1H4D; H4/B; H4FB; HIST1H4E; H4/J; H4FJ; HIST1H4F; H4/C; H4FC; HIST1H4H; H4/H; H4FH; HIST1H4I; H4/M; H4FM; HIST1H4J; HIST4/E; H4FE; HIST1H4K; H4/D; H4FD; HIST1H4L; H4/K; H4FK; HIST2H4A; H4/N; H4F2; H4FN; HIST2H4; HIST2H4B; H4
Gene ID (Entrez) NP_001029249
Format Affinity Purified
Storage Requirements 2-8°C for 12 months
Purification Method Affinity purified