Please login to your online account to display your discounted pricing

anti-dimethyl Histone H3 (Lys4), Polyclonal, EMD Millipore™


Manufacturer: emd millipore corporation  ABE250S

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. ABE250SMI

  • / Each
 

Description & Specifications

Specifications

Antigen dimethyl Histone H3 (Lys4)
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding Histone H3 methylated at Lys4.
Purification Method Affinity purified
Quantity 10μL
Storage Requirements 2-8°C for 12 months
Primary or Secondary Prrimary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) NP_003484.1
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Rat