anti-Dimethyl Histone H3 (Lys4), Clone: CMA303, EMD Millipore™
Manufacturer:  Millipore Sigma 051338
 View more versions of this product
Catalog No. 05-133-8MI
Specifications
Specifications
Dimethyl Histone H3 (Lys4)
CMA303
Purified mouse monoclonal IgG1κ in PBS with 0.05% sodium azide.
Purified
Murine
IgG1, κ
100μg
Primary
HumanþVertebrate
Chromatin ImmunoprecipitationþDot BlotþELISAþImmunocytochemistryþImmunoprecipitationþWestern Blot
Unconjugated
NM_003493
H3.4, H3/g , H3/t , H3FT , H3T, H3t , MGC126886, MGC126888 , OTTHUMP00000037945
Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, dimethylated on Lys4, conjugated to KLH.
Protein G purfied
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal