Please login to your online account to display your discounted pricing

anti-Dimethyl Histone H3 (Lys4), Clone: CMA303, EMD Millipore™


Manufacturer: emd millipore corporation  051338S

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 05-133-8SMI

  • / Each
 

Description & Specifications

Specifications

Immunogen Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, dimethylated on Lys4, conjugated to KLH.
Isotype κ
Gene Symbols H3.4, H3/g , H3/t , H3FT , H3T, H3t , MGC126886, MGC126888 , OTTHUMP00000037945
Storage Requirements 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications ELISA
Applications Immunocytochemistry
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Vertebrate
Antigen Dimethyl Histone H3 (Lys4)
Clone CMA303
Formulation Purified mouse monoclonal IgG1κ in PBS with 0.05% sodium azide.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Format Purified
Purification Method Protein G purfied
Quantity 10μg
Gene Accession No. NM_003493
Host Species Mouse