Please login to your online account to display your discounted pricing

anti-dimethyl Histone H3 (Arg26), Asymmetric, Polyclonal, EMD Millipore™


Manufacturer: emd millipore corporation  ABE411S

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. ABE411SMI

  • / Each
 

Description & Specifications

Specifications

Antigen dimethyl Histone H3 (Arg26), Asymmetric
Format Affinity Purified
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Applications Dot Blot
Applications Immunocytochemistry
Applications Western Blotting
Purification Method Affinity purified
Storage Requirements 2-8°C for 12 months
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ
Immunogen KLH-conjugated linear peptide corresponding to a region near the N-terminus of human Histone H3
Quantity 10μg
Primary or Secondary Prrimary
Gene ID (Entrez) NP_003520
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal