Please login to your online account to display your discounted pricing

anti-Dicer, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 1423-1463 of human Dicer protein were used as the immunogen for this antibody.
Antigen Dicer
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223891

 
novus biologicals canada
NBP223891
Unlabeled 0.1mL Each for $309.05

NBP223891SS

 
novus biologicals canada
NBP223891SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 1423-1463 of human Dicer protein were used as the immunogen for this antibody.
Antigen Dicer
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Gene Accession No. Q9UPY3
Gene ID (Entrez) 23405
Formulation 100μL of antibody; crude rabbit sera with 50% glycerol