missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Dicer, Polyclonal, Novus Biologicals™

$109.00 - $362.00

Specifications

Host Species Rabbit
Immunogen Rabbit anti-mouse Dicer polyclonal antibody was raised against a peptide corresponding to amino acids 132-150 and 1017-1033 of mouse Dicer (EDL18787).
Antigen Dicer
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB200591SS Novus Biologicals Canada
NB200591SS
0.025mg Each for $109.00
NB200591 Novus Biologicals Canada
NB200591
0.1mg Each for $362.00
Specifications Description & Specifications

Specifications

Rabbit
Dicer
Purified
RUO
In mouse ovary, a 217 kDa band is observed. Cross-reacts with Mouse. Expected to cross-react with Human and Rat due to over 90% sequence identity with immunogen, but testing shows no response in human and rat ovary at the recommended testing conditions. Not yet tested in other species.
PBS containing 0.05% BSA
Rabbit anti-mouse Dicer polyclonal antibody was raised against a peptide corresponding to amino acids 132-150 and 1017-1033 of mouse Dicer (EDL18787).
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
23405
0.5mg/mL