Please login to your online account to display your discounted pricing

anti-Dicer, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200591

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200591

  • / Each
 

Description & Specifications

Specifications

Antigen Dicer
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Immunogen Rabbit anti-mouse Dicer polyclonal antibody was raised against a peptide corresponding to amino acids 132-150 and 1017-1033 of mouse Dicer (EDL18787).
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23405
Test Specificity In mouse ovary, a 217 kDa band is observed. Cross-reacts with Mouse. Expected to cross-react with Human and Rat due to over 90% sequence identity with immunogen, but testing shows no response in human and rat ovary at the recommended testing conditions. Not yet tested in other species.
Target Species Human
Target Species Murine