Please login to your online account to display your discounted pricing

anti-DGAT2, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human DGAT2 protein (within residues 200-300). [Swiss-Prot Q96PD7]
Antigen DGAT2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP171701SS

 
novus biologicals canada
NBP171701SS
Unlabeled 0.025mL Each for $109.00

NBP171701

 
novus biologicals canada
NBP171701
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human DGAT2 protein (within residues 200-300). [Swiss-Prot Q96PD7]
Antigen DGAT2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human and mouse. Immunogen has 100% identity to rat, bovine and porcine, and 88% identity to Xenopus.
Gene Accession No. Q96PD7
Gene ID (Entrez) 84649
Formulation PBS, 30% glycerol
Concentration 1mg/mL