Please login to your online account to display your discounted pricing

anti-DGAT2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP171700SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP171700SS

  • / Each
 

Description & Specifications

Specifications

Antigen DGAT2
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Yeast
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q96PD7
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human DGAT2 protein (within residues 100-200). [Swiss-Prot Q96PD7]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 84649
Test Specificity Human, yeast and mouse. Immunogen has 100% identity to rat, bovine and porcine, and 86% identity to Xenopus.
Target Species Human
Target Species Murine
Target Species Yeast