Please login to your online account to display your discounted pricing

anti-DEPTOR/DEPDC6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227209SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227209SS

  • / Each
 

Description & Specifications

Specifications

Antigen DEPTOR/DEPDC6
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. Q8TB45
Host Species Rabbit
Immunogen A portion of amino acids 210-260 from human DEPTOR was used as the immunogen for this antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 64798
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, panda, new world monkey, dog, horse, macaque, rhesus monkey, white-cheeked gibbon, orangutan, rat, european pole-cat, and chimpanzee, 95% in cow, african bush elephant, opossum, mouse, rabbit, and pig, 74% in Carolina anole, chicken, wild turkey, zebra finch, and western clawed frog, 68% in zebrafish, and 63% homologous in guinea pig, mole rat, and cichlid fish.
Target Species Canine
Target Species Equine
Target Species Feline
Target Species Human
Target Species Primate
Target Species Rat