Please login to your online account to display your discounted pricing

anti-DEPTOR/DEPDC6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227209

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227209

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 64798
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q8TB45
Quantity 0.1mg
Antigen DEPTOR/DEPDC6
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, panda, new world monkey, dog, horse, macaque, rhesus monkey, white-cheeked gibbon, orangutan, rat, european pole-cat, and chimpanzee, 95% in cow, african bush elephant, opossum, mouse, rabbit, and pig, 74% in Carolina anole, chicken, wild turkey, zebra finch, and western clawed frog, 68% in zebrafish, and 63% homologous in guinea pig, mole rat, and cichlid fish.
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Format Purified
Regulatory Status RUO
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rat
Immunogen A portion of amino acids 210-260 from human DEPTOR was used as the immunogen for this antibody.
Host Species Rabbit
Primary or Secondary Primary
Applications Western Blotting