Please login to your online account to display your discounted pricing

anti-DEPTOR/DEPDC6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227083

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227083

  • / Each
 

Description & Specifications

Specifications

Antigen DEPTOR/DEPDC6
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Gene Accession No. Q8TB45
Host Species Rabbit
Immunogen A portion of amino acids 230-300 of human DEPOR/DEPDC6 was used as the immunogen. 243 258
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 64798
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee, horse and rat 93% homologous in mouse andcow, and 87% homologous in chicken.
Target Species Equine
Target Species Human
Target Species Primate
Target Species Rat