missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-DEPTOR/DEPDC6, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 210-260 from human DEPTOR was used as the immunogen for this antibody.
Antigen DEPTOR/DEPDC6
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227209 Novus Biologicals Canada
NBP227209
0.1mg Each for $255.80
NBP227209SS Novus Biologicals Canada
NBP227209SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
DEPTOR/DEPDC6
Purified
RUO
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, panda, new world monkey, dog, horse, macaque, rhesus monkey, white-cheeked gibbon, orangutan, rat, european pole-cat, and chimpanzee, 95% in cow, african bush elephant, opossum, mouse, rabbit, and pig, 74% in Carolina anole, chicken, wild turkey, zebra finch, and western clawed frog, 68% in zebrafish, and 63% homologous in guinea pig, mole rat, and cichlid fish.
64798
0.5mg/mL
A portion of amino acids 210-260 from human DEPTOR was used as the immunogen for this antibody.
Polyclonal
Primary
Western Blot
Q8TB45
0.1mg antibody in 0.2mL PBS containing 0.05% BSA.