missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-DEDD2, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 175-193 (APAAPQQQSEPARPSSEGK) of human DEDD2/Flame-3 was used as immunogen; GenBank no. gi|19923050|ref|NP_579874.1|
Antigen DEDD2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056138T Novus Biologicals Canada
NB10056138SS
0.025mL Each for $144.63
NB10056138 Novus Biologicals Canada
NB10056138
0.05mL Each for $222.95
Specifications Description & Specifications

Specifications

Rabbit
DEDD2
Antisera
RUO
DEDD2 .
PBS
A synthetic peptide corresponding to amino acids 175-193 (APAAPQQQSEPARPSSEGK) of human DEDD2/Flame-3 was used as immunogen; GenBank no. gi|19923050|ref|NP_579874.1|
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
162989