Please login to your online account to display your discounted pricing

anti-DEC2/SHARP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP119613SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP119613SS

  • / Each
 

Description & Specifications

Specifications

Antigen DEC2/SHARP1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS
Gene Accession No. Q9C0J9
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PAI1/Serpine 1 protein (between residues 1-75) [UniProt Q9C0J9]
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 79365
Test Specificity Human, Mouse, Rat. Immunogen has 100% homology to bovine and canine.
Target Species Human
Target Species Murine
Target Species Rat