Please login to your online account to display your discounted pricing

anti-DEC1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227151SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227151SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8553
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Primary or Secondary Primary
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, pig, new world monkey, panda, cow, dog, orangutan, rhesus monkey, white-cheeked gibbon, macaque and sheep. 89% in horse. 83% in opossum, chicken and platypus. 81% in zebrafish and zebra finch. 78% in lizard 69% in Atlantic salmon and pufferfish and 63% homologous in Japanese ricefish.
Quantity 0.025mg
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Regulatory Status RUO
Format Purified
Immunogen A portion of amino acids 20-88 of human Dec1 was used as the immunogen.
Host Species Rabbit
Antigen DEC1