Please login to your online account to display your discounted pricing

anti-DEC1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227151

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227151

  • / Each
 

Description & Specifications

Specifications

Antigen DEC1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Host Species Rabbit
Immunogen A portion of amino acids 20-88 of human Dec1 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8553
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, pig, new world monkey, panda, cow, dog, orangutan, rhesus monkey, white-cheeked gibbon, macaque and sheep. 89% in horse. 83% in opossum, chicken and platypus. 81% in zebrafish and zebra finch. 78% in lizard 69% in Atlantic salmon and pufferfish and 63% homologous in Japanese ricefish.
Target Species Human