Please login to your online account to display your discounted pricing

anti-DEC1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 20-88 of human Dec1 was used as the immunogen.
Antigen DEC1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227151SS

 
novus biologicals canada
NBP227151SS
Unlabeled 0.025mg Each for $109.00

NBP227151

 
novus biologicals canada
NBP227151
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 20-88 of human Dec1 was used as the immunogen.
Antigen DEC1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, pig, new world monkey, panda, cow, dog, orangutan, rhesus monkey, white-cheeked gibbon, macaque and sheep. 89% in horse. 83% in opossum, chicken and platypus. 81% in zebrafish and zebra finch. 78% in lizard 69% in Atlantic salmon and pufferfish and 63% homologous in Japanese ricefish.
Gene ID (Entrez) 8553
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL