Please login to your online account to display your discounted pricing

anti-DEC1, Polyclonal, Novus Biologicals™

$139.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503]
Antigen DEC1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001800

 
novus biologicals canada
NB1001800
Unlabeled 0.1mL Each for $384.00

NB1001800SS

 
novus biologicals canada
NB1001800SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503]
Antigen DEC1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Gene Accession No. O14503
Gene ID (Entrez) 8553
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL