Please login to your online account to display your discounted pricing

anti-DEC1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001800

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001800

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8553
Conjugate Unlabeled
Gene Accession No. O14503
Regulatory Status RUO
Concentration 1mg/mL
Formulation Tris-citrate/phosphate, pH 7-8
Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Quantity 0.1mL
Antigen DEC1
Primary or Secondary Primary
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503]
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal