Please login to your online account to display your discounted pricing

anti-DDX41, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225247SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225247SS

  • / Each
 

Description & Specifications

Specifications

Antigen DDX41
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg of DDX41 antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. NP_057306
Host Species Rabbit
Immunogen Within the range of amino acids 44-159 of human DDX41 were used as the immunogen for this antibody.
Isotype IgG
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51428
Target Species Human