Please login to your online account to display your discounted pricing

anti-DDX41, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225247

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225247

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 51428
Formulation 100μg of DDX41 antibody in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Concentration 0.5mg/mL
Gene Accession No. NP_057306
Immunogen Within the range of amino acids 44-159 of human DDX41 were used as the immunogen for this antibody.
Antigen DDX41
Host Species Rabbit
Format Purified
Applications Western Blotting
Isotype IgG
Cross Reactivity Human
Quantity 0.1mg
Regulatory Status RUO