missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-DDX41, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Within the range of amino acids 44-159 of human DDX41 were used as the immunogen for this antibody.
Antigen DDX41
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP225247 Novus Biologicals Canada
NBP225247
0.1mg Each for $307.00
NBP225247SS Novus Biologicals Canada
NBP225247SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Within the range of amino acids 44-159 of human DDX41 were used as the immunogen for this antibody.
Polyclonal
Primary
Western Blot
51428
0.5mg/mL
IgG
DDX41
Purified
RUO
NP_057306
100μg of DDX41 antibody in 200μL PBS containing 0.05% BSA