Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227408

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227408

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 30835
Monoclonal or Polyclonal Polyclonal
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, ape, and orangutan, 94% in old World monkey, rhesus monkey, macaque, white-cheeked gibbon, and chimpanzee, 89% in rat, and 83% homologous in mouse.
Quantity 0.1mg
Antigen DC-SIGN/CD209
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Immunogen A portion of amino acids 320-370 from human DC-SIGN (CD209) was used as the immunogen for this antibody.
Regulatory Status RUO
Applications Western Blotting
Conjugate Unlabeled
Primary or Secondary Primary
Host Species Rabbit