Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 320-370 from human DC-SIGN (CD209) was used as the immunogen for this antibody.
Antigen DC-SIGN/CD209
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227408

 
novus biologicals canada
NBP227408
Unlabeled 0.1mg Each for $255.80

NBP227408SS

 
novus biologicals canada
NBP227408SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 320-370 from human DC-SIGN (CD209) was used as the immunogen for this antibody.
Antigen DC-SIGN/CD209
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, ape, and orangutan, 94% in old World monkey, rhesus monkey, macaque, white-cheeked gibbon, and chimpanzee, 89% in rat, and 83% homologous in mouse.
Gene ID (Entrez) 30835
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL