Please login to your online account to display your discounted pricing

anti-DAZL, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 50-100 of mouse DAZL was used as the immunogen.
Antigen DAZL
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223663

 
novus biologicals canada
NBP223663
Unlabeled 0.1mL Each for $257.41

NBP223663SS

 
novus biologicals canada
NBP223663SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 50-100 of mouse DAZL was used as the immunogen.
Antigen DAZL
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a and CRA_c), monkey, 81% homologous in cow and rat, and 78% homologous in mouse. Based on amino acid content, the expected approximate size of FOXC1 is 60kD. Market data (including siRNA knockdown of FOXC1) and blocking datasuggests that the observed size of endogenous FOXC1is close to 48kD.
Gene Accession No. Q92904
Gene ID (Entrez) 1618
Formulation 100μL of crude rabbit sera with 50% glycerol