Please login to your online account to display your discounted pricing

anti-DAZL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223663SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223663SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1618
Formulation 100μL of crude rabbit sera with 50% glycerol
Gene Accession No. Q92904
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Immunogen A portion of amino acid 50-100 of mouse DAZL was used as the immunogen.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a and CRA_c), monkey, 81% homologous in cow and rat, and 78% homologous in mouse. Based on amino acid content, the expected approximate size of FOXC1 is 60kD. Market data (including siRNA knockdown of FOXC1) and blocking datasuggests that the observed size of endogenous FOXC1is close to 48kD.
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Host Species Rabbit
Regulatory Status RUO
Format Antiserum
Quantity 0.025mL
Primary or Secondary Primary
Antigen DAZL