Please login to your online account to display your discounted pricing

anti-Daxx, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056136SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056136T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1616
Formulation PBS
Conjugate Unlabeled
Test Specificity DAXX . Reacts with Human, Canine, Bovine, Primate and rat. 95% sequence identity with mouse.
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide corresponding to amino acids 272-291 (ERLINKPGPDTFPDYGDVLR
Quantity 0.025mL
Format Antiserum
Regulatory Status RUO
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen Daxx