Please login to your online account to display your discounted pricing

anti-Dact1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 725-775 of human Dapper homolog 1 was used as the immunogen for the antibody.
Antigen Dact1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224768SS

 
novus biologicals canada
NBP224768SS
Unlabeled 0.025mg Each for $109.00

NBP224768

 
novus biologicals canada
NBP224768
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 725-775 of human Dapper homolog 1 was used as the immunogen for the antibody.
Antigen Dact1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Based on immunogon sequence homology (76%), the antibody may cross-react with Dact2 The amino acid sequence used as immunogen is 100% homologous in human, mouse, dog, chicken, chimpanzee, zebrafish and cow.
Gene ID (Entrez) 51339
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL